Would you like to log in again?
Microchem® by AlphaTec™ 68-CFR Flame Retardant Coveralls with Serged and Taped Seams, Ansell
Supplier: Ansell
RD96-T-92-111-09 RD96-T-92-111-07 RD96-T-92-111-05 RD96-T-92-111-04 RD96-T-92-111-06 RD96-T-92-111-03 RD96-T-92-111-08 RD96-T-92-111-02
10836-278CS 1201.77 USD InStock
10836-278 10836-276 10836-458 10836-456 10836-280 10836-462 10836-274 10836-460
Microchem® by AlphaTec™ 68-CFR Flame Retardant Coveralls with Serged and Taped Seams, Ansell
Disposable Garments
MICROCHEM® by AlphaTec™ CFR is a flame retardant and antistatic fabric offering protection from particulates and light liquid spray or splash without compromising worker protection in the event of a flash fire.
  • Flame retardant treated fabric with PVC barrier film offering wearers protection from liquid chemicals
  • Versatile in most applications where there is the need for protection from chemical spray without compromising wearer protection in the event of a flash fire
  • Optimized body fit improves wearer comfort and safety
  • Highly visible bright red color to improve worker safety
Must be worn over thermal protective garments, such as NOMEX®, and never to be worn next to the skin. These disposable latex coveralls feature an elasticated hood, wrists, and ankles with double zipper closure. Typically used in oil and petrochemical industries, petroleum distribution and processing, and utilities.
Certifications: Tested according to EN 1073-2, EN1149-5 / AATCC 76, EN 14126 / ASTM F1671, EN 14605 Type 3, EN 14605 Type 4, EN ISO 13982-1 Type 5, EN ISO 14116 INDEX 1/0/0, ANSI/ISEA 101.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty