Would you like to log in again?
Microchem® by AlphaTec™ 68-2000 Industrial Coveralls with Liquid Protection and Bound Seams, Ansell
Supplier: Ansell
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
WH20-B-92-107-07 WH20-B-92-103-06 WH20-B-92-111-09 WH20-B-92-111-05 WH20-B-92-111-04 WH20-B-92-111-02 WH20-B-92-103-03 WH20-B-92-103-04 WH20-B-92-103-08 WH20-B-92-111-03 WH20-B-92-111-08 WH20-B-92-107-06 WH20-B-92-107-05 WH20-B-92-103-02 WH20-B-92-111-06 WH20-B-92-107-08 WH20-B-92-103-07 WH20-B-92-111-07 WH20-B-92-107-09 WH20-B-92-107-03 WH20-B-92-107-04 WH20-B-92-103-05 WH20-B-92-103-09
76498-906CS 323.51 USD InStock
76498-906 10836-366 76498-918 76498-904 76498-916 10836-166 10836-364 76498-914 10836-358 76498-912 10836-368 76498-908 10836-370 10836-172 76498-910 10836-160 10836-170 10836-362 10836-164 10836-174 10836-372 10836-360 10836-162
Microchem® by AlphaTec™ 68-2000 Industrial Coveralls with Liquid Protection and Bound Seams, Ansell
Disposable Garments
MICROCHEM® by AlphaTec™ 2000 coveralls provide both protection and comfort with exceptional liquid and particulate protection
  • Excellent liquid penetration resistance and barrier to fine particulates
  • ASTM F1761 and EN 14126:2003 resistance to penetration of blood, body fluids, and blood-borne pathogens
  • Moisture vapor permeable to help reduce the risk of heat stress
  • Low linting reduces the risk of fiber contamination in some critical areas
  • Antistatic tested according to EN1149-5 & AATCC76 (8EMSL test method)
These disposable latex coveralls with bound seams have a 2-way front zipper with resealable storm flap and elasticated wrists, waist, and ankles.
Ideal for a wide range of industrial applications.
Certifications: Tested according to EN 14126 / ASTM F1671, DIN 32781, EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5B, ISO 17491-4 Type 6B.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty