Would you like to log in again?
Microchem® by AlphaTec™ 68-2300 Lightweight Chemical Protection Coveralls with Bound Seams, Ansell
Supplier: Ansell
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
AlphaTec®
YY23-B-92-111-02 YY23-B-92-111-04 YY23-B-92-103-08 YY23-B-92-103-03 YY23B9214706 YY23-B-92-103-04 YY23-B-92-111-07 YY23-B-92-111-05 YY23-B-92-111-08 YY23-B-92-103-07 YY23-B-92-103-02 YY23-B-92-103-06 YY23-B-92-111-06 YY23-B-92-103-09 YY23-B-92-111-03 YY23-B-92-111-09 YY23-B-92-103-05
10836-202CS 379.38 USD InStock
10836-202 10836-410 10836-200 10836-188 10836-206 10836-414 10836-204 10836-412 10836-418 10836-208 10836-416 10836-190 10836-194 10836-192 10836-198 10836-186 10836-196
Microchem® by AlphaTec™ 68-2300 Lightweight Chemical Protection Coveralls with Bound Seams, Ansell
Disposable Garments
MICROCHEM® by AlphaTec™ 2300 provides an excellent barrier to harmful chemicals and blood borne pathogens, while being lightweight and relatively strong and durable.
  • Lightweight yet strong and durable
  • Typical coverall features include respirator fit hood and zip flap with self-adhesive tape closure
  • Finger loops
  • Antistatic tested according to EN1149-5 and AATCC76
These disposable latex coveralls provide a barrier to numerous inorganic liquid chemicals including acids and bases. They achieve the highest classifications for protection from biological agents in accordance with EN 14126:2003 and ASTM F1671 for penetration of blood, bodily fluids, and blood-borne pathogens. Coveralls feature an elasticated hood, waist, wrists, and ankles, along with a 2-way front zipper with resealable storm flap. These garments can be used in environmental clean-up, sewage purification installations, and composites manufacturing.
Certifications: Tested according to EN 1073-2, EN1149-5 / AATCC 76, EN 14126 / ASTM F1671, EN ISO 13982-1 Type 5-B, ISO 17491-4 Type 6-B, ANSI/ISEA 101.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty