Would you like to log in again?
Microchem® by AlphaTec™, 68-5000 Chemical Protection Coveralls with Hood and Ultrasonically Welded and Taped Seams, Ansell
Supplier: Ansell
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
OR50-T-92-111-04 OR50-T-92-111-08 OR50T9212208 OR50-T-92-111-09 OR50-T-92-111-03 OR50-T-92-111-07 OR50-T-92-111-02 OR50-T-92-122-09 OR50-T-92-111-05 OR50-T-92-111-06
10836-444CS 459.72 USD InStock
10836-444 10836-256 10836-454 10836-452 10836-254 10836-448 10836-270 10836-252 10836-272 10836-250
Microchem® by AlphaTec™, 68-5000 Chemical Protection Coveralls with Hood and Ultrasonically Welded and Taped Seams, Ansell
Disposable Garments
MICROCHEM® by AlphaTec™ 5000 reaches new levels in chemical protection and has been engineered to protect. The highly visible multi-layer fabric is strong, durable, and suitable for workers in extremely hazardous areas, including HAZMAT response teams.
  • Barrier to numerous organic and inorganic chemicals and biological hazards
  • Multi-layer material, which is lightweight yet strong and durable
  • Innovative design features include liquid-tight dual zip closure without the need for additional taping
  • Bright orange color for improved worker safety
Breakthrough time against 14 of 15 chemicals listed in EN ISO 6529 >480 minutes
Typical areas of application include industrial and tank cleaning, sewage purification installations, chemicals, oil and petrochemicals, mining, agriculture, HAZMAT emergency response (i.e. level B), first response, and fire service.
Coveralls feature a 2-piece hood, double zip closure, and double cuffs with a knitted inner cuff.
Certifications: Tested according to EN 1073-2, EN1149-5 / AATCC 76, EN 14126 / ASTM F1671, EN 14605 Type 3-B, EN 14605 Type 4-B, EN ISO 13982-1 Type 5-B, ANSI/ISEA 101.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty