Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Universal® Premium Blue Masking Tape with Bloc-it™ Technology, Essendant LLC MS
Supplier: Essendant
Universal®
UNVPT14049 UNVPT14019 UNVPT14025
102962-956PK 15.38 USD InStock
102962-956 10836-646 102962-958
Universal® Premium Blue Masking Tape with Bloc-it™ Technology, Essendant LLC MS
Tapes
Premium blue masking tape.
  • Provides clean paint lines and removes cleanly and easily
  • Excellent conformability as well as smooth backing for superior paint lines and reduced fingertip abrasion
  • 14-day UV-resistant
  • Consistent, easy unwind and resistant to humidity and paint bleed-through
  • Specialty grade
  • Crepe paper finish
Blue fine crepe paper masking tape with Bloc-it™ Clean Line Technology is a specially formulated pressure-sensitive, synthetic rubber adhesive designed for exposure to sunlight for up to fourteen days. Exhibits easy, clean removal from most surfaces. Also has secure adhesion and is stain-resistant. Synthetic rubber adhesive material. Dispenser not included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty