Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Liquid Disposal Safety Cans, Justrite®
Supplier: Justrite
14265 14565
10836-904EA 421.65 USD InStock
10836-904 10836-902
Liquid Disposal Safety Cans, Justrite®
Safety Cans
These high-density polyethylene (HDPE) disposal cans with built-in fill gauge provide superior chemical resistance and leak protection.
  • Superior chemical resistance and leak protection
  • Spring-loaded sealed cap automatically vents
  • Stainless steel flame arrester
  • Design resists accidental tip over
Built-in polyethylene fill gauge indicator pops up to warn you that the container is almost full.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty