Would you like to log in again?
Biohazard Waste Cans, Justrite®
Supplier: Justrite
5915 05910R 5914 05930R 05935 5934
10836-828EA 258.81 USD InStock
10836-828 10836-916 10836-824 10836-912 10836-826 10836-914
Biohazard Waste Cans, Justrite®
Bins Waste Bins
Leakproof waste cans with self-closing lids provide safe, temporary collection of biohazardous waste.
  • Great for medical facilities and research labs
  • Foot-operated, self-closing lid reduces risk of contamination
  • Reduce closure noise with SoundGard™ models
  • Sturdy design with steel construction
Safely contain biohazardous materials, including bloodborne pathogens and other potentially infectious materials (no sharps).
Meets OSHA 29 CFR 1910.1030 for the segregation of infectious waste and FM approved for use with alcohols and other flammable liquids.
Certifications: FM, TUV, OSHA.
Ordering information: Includes OSHA biohazard label on front; use optional label kit 25880 for additional labels to add to the sides, to meet California Health and Safety Code.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty