Would you like to log in again?
OHM Metrics™ CM400 Series Constant Monitor, Transforming Technologies
Supplier: Transforming Technologies
CM410PV CM420PV CM410 CM420 CM400
10837-024EA 134.84 USD InStock
10837-024 10837-022 10837-028 10837-026 10837-030
OHM Metrics™ CM400 Series Constant Monitor, Transforming Technologies
Static Control Monitors
The OHM Metrics™ CM400 Series single wire constant monitors are the most cost effective method of continuously testing the connection to ground of a person, wrist band, and coil cord to ensure ESD protection
Compatible with any standard single wire wrist strap
Easy to use: Simply plug in the wrist strap
Easy to install: Attach the monitor to the bench with included hardware, and plug it in
Constant monitors eliminate the need to test wrist straps and reduce ESD damage from broken wrist straps that go unnoticed. The systems are compatible with most single wire wrist straps.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty