Would you like to log in again?
Overhead Three Fan Ionizer, Transforming Technologies
Supplier: Transforming-Technologies
BFN803
10837-090EA 1737.16 USD InStock
10837-090
Overhead Three Fan Ionizer, Transforming Technologies
Ionizers
The BFN 802 ESD Overhead ionizing blowers, now with LED Light technology, combine excellent ESD performance with user friendly features to create the premier Overhead ESD workstation solution.
The ionizers are equipped with energy efficient LED task lights, to help alleviate eye strain, headaches and even have a positive effect on mood - all while saving on energy costs
BFN series AC ionizers from Transforming Technologies create a dense and well-balanced ionization current. They are unique in their ability to deliver fast decay times with low offset voltages. Continuous balance and decay protection is assured by the reliable AC design. An integrated emitter point cleaner, removable front and rear fan guards, safety switch
combine to make the BFN 801 a user friendly ionizer.
Integrated emitter point cleaners, removable front and rear fan guards, and safety switches provide for safe, and easy maintenance access makes the BFN Overhead Ionizers very user friendly.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty