Would you like to log in again?
Ptec™ Airflex Ionizer, Transforming Technologies
Supplier: Transforming-Technologies
IN4000PE FL0020 IN4000
10837-122EA 2287.08 USD InStock
10837-122 10837-118 10837-096
Ptec™ Airflex Ionizer, Transforming Technologies
Ionizers
Ptec™ ionizers save time and money because they require no calibration
The IN4000 features the inherently stable output of all Ptec™ Ionizers and high frequency AC (68KHz) ion emission. The flexible metal neck, with multiple nozzle options, allows for easy operation and service of emitters. A foot switch and .01 micron filter combine to deliver clean air at the correct pressure when needed.
The FL0020 micro filter features a hollow fiber membrane that provides excellent filtration and has a long service life. The compact design is easy to install and comes with quick release connections for fast replacement. All materials used are compatible for the most stringent applications.
Emitter life is increased and maintenance frequency is reduced, due to 68KHz AC ion emission. The IN4000 flexible nozzle neck frees the operator's hands during assembly and manufacturing processes. A foot pedal controls both ionization and airflow, which reduces compressed air costs and extends the life of the ionizer. The model IN4000 ionizing air nozzle produces a 68KHz AC output of approximately 2200V and a continuous stream of balanced air ions.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty