Synonyms:
Dihydrogen oxideIce
WIR-USP-2XL
WIR-USP-1X6
Z0304
Z0304
WIR-USP-1X6
WIR-USP-20L
WIR-USP-20L
WIR-USP-2XL
WIR-USP-5XM
WIR-USP-5XM
10837-200EA
1680
USD
InStock
10837-200
CA10837-198
CA10837-196
10837-198
76353-870
CA10837-200
76312-764
CA76312-764
10837-196
CA76353-870
Water USP for irrigation (WIR), sterile
Water
Water for Irrigation, USP grade is a sterile, hypotonic, non-pyrogenic fluid or solvent. It is prepared by terminal distillation and contains no antimicrobial or bacteriostatic agents or added buffers. The pH is 5.0-7.0 and complies with the USP monograph for water for irrigation.
Manufactured under cGMP and ISO 13485 guidelines in an FDA registered facility.
An excellent choice for upstream and downstream media/buffer preparation in biopharmaceutical manufacturing applications. It is also widely used in medical device manufacturing/processing applications where a high quality, sterile water is required