Would you like to log in again?
COVERALLS WHITE XL 46-50 CS24
Supplier: KIMBERLY CLARK CORP
KLEENGUARD* Select Coveralls^ X-Large, white. Made from breathable MicroForce* Barrier SMS fabric. Protects against submicron sized particles (0.26um) and light liquid sprays. Abrasion-resistant. Plain coverall has zipper front and elastic back, wrists, and ankles. Meets 16 CFR 1610.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COVERALLS WHITE XL 46-50 CS24 10837-324 49104 Case of 24 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty