Would you like to log in again?
KleenGuard™ A60 Bloodborne Pathogen and Chemical Splash Protection Coveralls, Kimberly-Clark Professional
Supplier: Kimberly-Clark
KleenGuard®
45023 45094 45022-75 45027 45003 45024 45005 45097 45006 45092 45093 45095 45006 45092 45004 45025 45097-20 45003 45095 45232 45093-20 45024-75 45022 45007 45025-75 45023-75 45235-00 45096 45027-75 45096 45232 45002 45005 45094-20 45007 45235 45026
82003-928CS 560.02 USD InStock
CA82003-914 82003-928 CA82003-934 82003-926 CA82003-912 CA82003-918 82004-870 CA82003-916 37001-138 10837-390 37001-136 37001-134 82003-924 CA82003-932 37001-132 CA82003-930 82003-922 CA82004-870 CA82004-854 CA82003-926 82003-916 CA82003-924 82003-914 CA82003-928 CA37001-138 82003-918 82003-930 CA37001-136 82003-912 CA82003-922 CA37001-132 82003-934 82003-932 CA37001-130 CA82003-920 CA82004-848 82004-854
KleenGuard™ A60 Bloodborne Pathogen and Chemical Splash Protection Coveralls, Kimberly-Clark Professional
Disposable Garments
KleenGuard™ A60 coveralls are manufactured from a three-layer fabric.
  • Seamless front ensures increased protection in the primary exposure areas
  • Patented REFLEX™ design is 7.5 x less likely to rip out than ANSI minimums
  • Provides 12% more chest room and 6% longer body length for a more comfortable fit
  • Zipper front
Coveralls are ideally suited for use in emergency medical response, crime scene cleanup, embalming/forensics, tank cleaning, fiberglass manufacturing, liquid handling, and animal research applications.
The outer fabric layers are cloth-like, tear- and abrasion-resistant spunbond polypropylene designed for extended use. The inner layers are constructed of a breathable microporous film that provides resistance to numerous liquids and dry particulates for user protection, while allowing heat and sweat vapors to escape for increased comfort.
Certifications: Coveralls pass ASTM F1670/1671 testing for penetration of body fluids and blood-borne pathogens. Passes NFPA 99 for antistatic materials.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty