Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ultra Cyano HPLC Prep Columns, Restek

Supplier: Restek
9106559 9106519 9106579 9106517 9106568 9106518 9106558 9106569 9106578 9106557 9106567
10837-552EA 1332.83 USD InStock
10837-552 10837-556 10837-534 10837-554 10837-538 10839-258 10839-266 10837-536 10839-256 10839-264 10837-560
Ultra Cyano HPLC Prep Columns, Restek
Chromatography Columns
The Restek Cyano is a traditional monomeric cyanopropylsilane that is recommended for assays where alternate selectivity, or confirmation, to a C18 or C8 column is desired.

  • Particle size and pore size: 5 µm, spherical and 100 Å
  • Carbon load: 8%
  • End-cap: yes
  • pH range: 2.5 to 8
  • Temperature limit: 80 °C
  • USP phase code: L10
  • Phase category: cyano
  • Ligand type: cyanopropylsilane

It can be used in normal-phase, reversed-phase (best with mobile phase pH between 5 and 7), and HILIC modes. It is an excellent choice for the analysis of protonated bases. Easy scale-up from Restek analytical columns.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty