Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Viva C8 Columns, Restek

Supplier: RESTEK CORP
9513562-700 9513525-700 9513515-700 9513523-700 951356E 9513552 9513532 9513511 951351E 9513512-700 9513535-700 9513561 9513512 9513571 9513575-700 9513572-700 9513515 9513553-700 951355E 9513555 9513552-700 9513551 9513522-700 9513533-700 9513565-700 9513525 9513535 9513522 9513532-700 951357E 9513521 951352E 9513555-700 9513531 9513573-700 9513563-700 9513513-700 9513572 951353E
10838-792EA 672.76 USD InStock
10838-792 10838-772 10838-794 10838-796 10838-774 10838-776 10838-798 10838-810 10838-778 10838-812 10838-814 10838-816 10838-818 10840-098 10837-788 10837-766 10837-764 10837-768 10838-780 10838-782 10838-784 10838-786 10838-788 10838-800 10838-802 10838-804 10837-780 10838-806 10837-772 10838-808 10837-770 10837-776 10837-798 10837-796 10837-774 10837-778 10840-100 10840-102 10838-790
Viva C8 Columns, Restek
Chromatography Columns
Restek's C8 is a conventional monomeric octylsilane column offering a shorter alkyl chain to provide less hydrophobic retention and improved basic peak shape over a traditional C18 phase.

  • Rugged, spherical particles with 300 Å pore size
  • Particlei size and pore size: 5 µm, spherica and 300 Å
  • Carbon load: 5%
  • End-cap: yes
  • pH range and temperature limit: 2.5 to 8 and 80 °C
  • USP phase code: L7
  • Phase category and Ligand type: C8, octylsilane and monomeric C8

Like Restek's C18, this general-purpose Restek C8 is suitable for a wide range of compounds from acidic through slightly basic.

Restek's C8 has high proportion of pore/surface area available to large molecules and excellent for separating peptides or proteins.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty