Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ultra C4 Columns, Restek

Supplier: Restek
9102562 9102355 9102552-700 9102332-700 9102335 9102575-700 9102335-700 9102312-700 9102315-700 9102535 9102362-700 9102552 9102365 9102313-700 9102362 910236E 9102355-700 9102532 9102515-700 9102522 9102565-700 910231E 9102572-700 9102555-700 9102363-700 9102512-700 9102332 910256E 9102352-700 910255E 910257E 9102573-700 9102365-700 9102562-700 9102555 9102525 9102563-700 9102353-700 9102565 910251E 910235E 9102553-700 9102535-700 9102532-700 9102575 9102512 910233E 9102572 9102513-700 9102352 910253E 9102312 9102315 910252E 9102515 9102525-700 9102523-700 9102522-700
10839-238EA 733.72 USD InStock
10839-238 10839-236 10839-232 10839-196 10839-230 10837-982 10837-980 10837-986 10840-490 10837-984 10837-988 10839-226 10839-224 10839-220 10839-188 10837-992 10840-480 10839-194 10840-482 10837-990 10839-190 10837-996 10837-994 10840-488 10837-998 10840-484 10840-486 10838-010 10838-014 10839-218 10838-016 10838-018 10839-214 10839-212 10840-470 10839-184 10840-472 10839-182 10837-964 10837-962 10837-968 10840-478 10837-966 10840-474 10840-476 10838-000 10839-208 10838-002 10839-206 10838-004 10838-006 10839-202 10838-008 10839-200 10837-970 10837-974 10837-978 10837-976
Ultra C4 Columns, Restek
Chromatography Columns
Exceptionally stable C4 packing with high bonding coverage and base deactivation.

  • Particle size: 3 µm or 5 µm, spherical
  • Pore size: 100 Å
  • Carbon load: 9%
  • End-cap: yes
  • pH range and temperature limit: 2.5 to 8 and 80 °C
  • USP phase code: L26
  • Phase category and ligand type: C4, butylsilane and monomeric C4

These columns feature less retention than C18 or C8 phases.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty