Would you like to log in again?
Pinnacle® DB Silica HPLC Columns (USP L3), Restek
Supplier: Restek
9410512-700 9410565-700 941051E 9410525-700 941056E 9410535-700 9410332 9410212 9410555 9410355-700 9410313-700 9410365-700 9410335-700 9410352-700 9410333-700 9410553-700 9410555-700 9410312 9410312-700 9410335 9410513-700 9410252 9410363-700 941057E 9410315 9410512 941052E 9410332-700 9410515-700 941055E 9410575-700 9410552 9410565 9410362 9410562-700 941035E 9410353-700 9410362-700 9410532 9410232 9410573-700 941036E 9410562 941033E 9410575 9410365 9410552-700 9410535 9410315-700 9410525 9410515 9410572-700 9410352 9410355 941031E 941053E 9410563-700 9410532-700 9410572 9410522-700 9410533-700 9410523-700 9410522
10838-472EA 597.54 USD InStock
10838-472 10838-474 10838-476 10838-510 10059-836 10838-478 10838-512 10059-834 10838-514 10838-516 10838-518 10837-662 10837-622 10837-664 10837-620 10837-626 10838-480 10837-628 10838-482 10838-462 10838-464 10838-466 10838-500 10838-468 10838-502 10838-504 10838-506 10838-508 10837-598 10059-832 10837-638 10838-470 10838-494 10838-496 10838-452 10839-736 10838-454 10841-030 10838-498 10838-456 10838-458 10837-640 10837-644 10837-604 10837-602 10837-646 10837-608 10838-460 10838-484 10838-486 10838-520 10838-488 10838-522 10841-024 10841-028 10841-026 10837-652 10837-650 10837-610 10837-616 10837-614 10838-490 10838-492
Pinnacle® DB Silica HPLC Columns (USP L3), Restek
Chromatography Columns
Base-deactivated spherical silica is useful for normal-phase or HILIC separations.
  • For normal phase separations
  • Optimized for UHPLC stability
Pinnacle DB columns are 100% manufactured by Restek in our Bellefonte, Pennsylvania facility. Because performance begins with the support, our Pinnacle DB UHPLC columns start with base-deactivated spherical silica that is optimized for UHPLC stability. From there, we bond them with a wide variety of phases to give chromatographers a stable and selective column. Get the most out of your UHPLC system. Combine selectivity and efficiency by using Restek Pinnacle DB UHPLC columns.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty