Would you like to log in again?
ULTRA PFPP20 X 2.1MM 2-PK
Supplier: Avantor
Ultra PFP Propyl Guard Cartridge 5um^ Ultra PFP Propyl Guard Cartridge, Particle size: 5 um, Spherical, Size: 20 x 2.1 mm, Pore size: 100 Angstrom, Managing High Backpressure, Guard column requires Trident direct guard cartridge system
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
ULTRA PFPP20 X 2.1MM 2-PK 10840-280   Pack of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty