Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ultra Silica Columns, Restek

Supplier: Restek
9100515-700 9100353-700 9100535 910052E 910035E 9100315-700 910031E 9100572-700 9100312-700 910036E 9100335-700 910051E 9100522-700 9100365 9100573-700 9100362 9100515 9100565-700 9100555 9100352-700 9100575-700 9100362-700 9100355 9100513-700 910055E 9100332-700 9100525 9100365-700 9100522 9100535-700 9100532 9100335 9100553-700 9100555-700 9100355-700 910053E 9100332 910057E 9100313-700 9100525-700 9100552 9100562-700 9100512-700 910033E 910056E 9100572 9100563-700 9100552-700 9100532-700
10840-448EA 639.54 USD InStock
10840-448 10839-118 10839-078 10837-860 10840-450 10837-864 10837-862 10837-868 10840-412 10840-456 10837-866 10840-458 10840-414 10840-452 10839-068 10839-102 10839-064 10839-074 10837-872 10839-072 10837-870 10837-876 10837-874 10840-444 10840-446 10837-878 10840-440 10840-442 10840-426 10840-428 10839-098 10839-062 10837-880 10840-434 10840-436 10840-430 10840-432 10840-416 10840-418 10839-088 10839-086 10840-460 10839-094 10839-092 10840-466 10840-424 10840-468 10840-420 10840-464
Ultra Silica Columns, Restek
Chromatography Columns
Base-deactivated spherical silica is useful for normal-phase or HILIC separations.

  • Particle Size: 3 or 5 µm, spherical
  • Pore size: 100 Å
  • End-cap: no
  • pH range and temperature limit: 2.5 to 8 and 80 °C
  • USP phase code: L3
  • Phase category: bare silica
  • Ligand type: none

Excellent for separating peptides or proteins, these silica columns have rugged, spherical particles with a pore size of 100 Å. High proportion of pore/surface area available to large molecules.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty