Base-deactivated spherical silica is useful for normal-phase or HILIC separations.
- Particle Size: 3 or 5 µm, spherical
- Pore size: 100 Å
- End-cap: no
- pH range and temperature limit: 2.5 to 8 and 80 °C
- USP phase code: L3
- Phase category: bare silica
- Ligand type: none
Excellent for separating peptides or proteins, these silica columns have rugged, spherical particles with a pore size of 100 Å. High proportion of pore/surface area available to large molecules.