Would you like to log in again?
TraceGOLD™ TG-1MT Metal GC Columns
Supplier: Thermo Scientific
TraceGOLD
26M99-1300 26M99-3480 26M99-4120 26M99-1420 26M99-4130
10056-852EA 650.09 USD InStock
10056-852 75831-972 75814-690 10841-590 10056-854
TraceGOLD™ TG-1MT Metal GC Columns
Chromatography Columns
These non polar, metal columns allow you to work at temperatures up to 430 °C with no compromise in performance when analyzing highly active and aggressive compounds.
  • Nonpolar, 100% dimethyl polysiloxane
  • Low bleed for excellent signal-to-noise ratio, sensitivity and mass spectral integrity
  • Stable to 430 °C
These columns are recommended for hydrocarbons, GROs, drugs of abuse and PCBs.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty