Would you like to log in again?
GC Column, TG-BiodieselMT
Supplier: Thermo Scientific
26MB9-1932
10841-594EA 853.48 USD InStock
10841-594
GC Column, TG-BiodieselMT
Chromatography Columns
Used for glycerides analysis.
Certifications: ASTM D6584.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty