Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lactic acid ≥88%

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: 2-Hydroxypropionic acid2-Hydroxypropanoic acidDL-Lactic acid

L2016-500ML L2016-2.5LT
10841-788EA 123 USD InStock
10841-788 10841-786
Lactic acid ≥88%
Lactic acid
DL-Lactic Acid, Racemic, also known as milk acid, is used in various biochemical processes. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

GHS hazard : Corrosion
UN3265


Formula: H₃CCH(OH)COOH
MW: 90.08 g/mol
Density: 1,209
Storage Temperature: Ambient
MDL Number: MFCD00004520
CAS Number: 50-21-5
UN: 3265
ADR: 8,III
Order Now
Specification Test Results
Assay Approx. 88 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty