Synonyms:
(±)-3-Mercapto-1,2-dihydroxy propane
, (±)-3-Mercapto-1,2-propanediol
, (±)-α-Monothioglycerine
, alpha-Thioglycerol
, α-Monothioglycerol
, (±)-1-Thioglycerine
, 1-Thioglycerol
, Monothioglycerol
M1177-100ML
M1177-500ML
M1177-25ML
10841-882EA
228
USD
InStock
10841-882
10841-886
10841-884
3-Mercapto-1,2-propanediol ≥97%
3-Mercapto-1,2-propanediol
Monothioglycerol, also known as 3-Mercaptopropane-1,2-diol and thioglycerol, is often used as a matrix in liquid secondary ion mass spectrometry and fast atom bombardment mass spectrometry.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.