Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phosphorus, red, amorphous fine powder (DEA List I Chemical)

Supplier: SPECTRUM CHEMICAL MFG CORP

P1110-25GM P1110-100GM P1110-6X500GM P1110-500GM
10841-956CS 502 USD InStock
10841-956 10841-954 10841-952 10841-950
Phosphorus, red, amorphous fine powder (DEA List I Chemical)
Phosphorus
Phosphorus, Red (DEA List I Chemical), Amorphous, Fine Powder, a chemical element, is widely used in fertilizers as an essential plant nutrient. It is also used to make organophosphorus compounds. The United States Drug Enforcement Agency (DEA) maintains a list of illicit drugs and the chemical materials used to synthetically produce them. List I chemicals are those that are used in the manufacture of the controlled substances and are important to the manufacture of the substances. Regulatory forms are required to purchase this material

GHS hazard : Flame;Exclamation mark
UN1338


Formula: P
MW: 30.97 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00133771
CAS Number: 7723-14-0
UN: 1338
ADR: 4.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty