Would you like to log in again?
InLab® Sensors and Cables, METTLER TOLEDO
Supplier: Mettler-Toledo
30281924 30281897 30281926 30244732 30281913 30281928 30281899 30244733 30281912 30281927 30281898 30281896 30249570 30248828 30281925 30281918 30281913 30281915 30281918 30281911 30248120 30281916 30281920 30281912 30281910 30248112 30248120 30281921 30247850 30281899 30253098 30281920 30249570 30281923 30253098 30281928 30281916 30281911 30281915 30248832 30248830 30281910 30281923 30244733 30281921 30248826 30281896 30248832 30248826 30281924 30281917 30281898 30248112 30281919 30281922 30244732 30248830 30281922 30281926 30281919 30281925 30281917 30281927 30281897 30247850 30248828
10842-612EA 86.75 USD InStock
CA10842-620 10842-612 CA10842-622 10842-614 CA10842-624 10842-616 CA10842-626 10842-618 CA10842-628 10842-580 10842-582 CA10842-592 CA10842-594 CA10842-596 10842-620 CA10842-630 CA10842-598 10842-622 CA10842-632 10842-624 CA10842-634 10842-626 CA10842-636 10842-628 CA10842-638 10842-592 10842-594 10842-596 10842-630 10842-598 CA10842-640 10842-632 CA10842-642 10842-634 CA10842-600 10842-636 CA10842-646 CA10842-602 10842-638 CA10842-648 CA10842-604 CA10842-606 CA10842-608 10842-640 CA10842-650 10842-642 CA10842-652 10842-600 CA10842-610 CA10842-654 10842-602 10842-646 CA10842-612 10842-604 10842-648 CA10842-614 10842-606 CA10842-616 10842-608 CA10842-618 CA10842-580 CA10842-582 10842-650 10842-652 10842-610 10842-654
InLab® Sensors and Cables, METTLER TOLEDO
Electrodes pH Electrodes
METTLER TOLEDO electrodes combine innovative technologies for fast, accurate and reliable measurements.
  • Easy to use
  • Built to last
  • Performance to trust
All sensors are shipped ready to use
The well-designed InLab sensor construction, as well as the high-quality materials used for manufacturing guarantee a robust sensor - even in harsh applications. The versatile sensor portfolio ensures the prefectly suited sensor for every application, a match that will greatly lengthen the lifetime of the product.
Quality is more than a promise for us. To guarantee it, we test every single sensor. Only sensors which successfully pass the strict final product inspection receive an individual quality certificate, and are ready for delivery.
Everything you need to measure comes together with the sensor. Your work in the lab will be simplified by the spill-proof wetting cap, the automatic sensor recognition of ISM sensors, and the application-specificity of the sensor you select.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty