Would you like to log in again?
HOOD SIZE XSMALL
Supplier: ALSICO USA
Pullover Hoods^ Complete head coverage. Extra-long shoulder drape fits well into coverall or frock, preventing pullout. Elastic take-up in back with horizontal and vertical snaps for adjustment. Sewn-in replaceable over the nose 2-ply poly tricot facial. Color: White. Size: X-Small.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
HOOD SIZE XSMALL 10842-896 HB315887SPXS Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty