Would you like to log in again?
VWR® Disposable Coveralls with PVC Bootcovers and Bonded Seams
Supplier: VWR
VWR®
SQCB-001 SQCB-003 SQCB-004 SQCB-002 SQCB-006
10847-228CS 414.95 USD InStock
10847-228 10847-224 10847-222 10847-294 10847-292
VWR® Disposable Coveralls with PVC Bootcovers and Bonded Seams
Disposable Garments
Disposable coveralls are manufactured from 60gsm SF fabric, and they feature 250gsm nonskid PVC sole boots covers.
  • Bonded seams
  • Low particle shielding
  • Comfortable design
All coveralls feature bonded seams, with materials and construction that ensure low particle shielding. All garments are easy to wear.
Certifications: ISO9001, ISO13485, ISO14001, CE, GMP.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty