Would you like to log in again?
FlexTech™ Seamless Knit Palm Coated Glove with Impact Pads, Sandy Nitrile Palm, Wells Lamont®
Supplier: Wells Lamont
FlexTech™
I2449TXXL I2449TM I2449TL I2449TS I2449TXL
10847-258PK 480.03 USD InStock
10847-258 10847-256 10847-264 10847-262 10847-260
FlexTech™ Seamless Knit Palm Coated Glove with Impact Pads, Sandy Nitrile Palm, Wells Lamont®
Gloves
The machine-knit shell of these gloves provides comfort and dexterity.
  • Machine knit shell for comfort and dexterity
  • Work best in oil and gas, mining, and heavy construction applications
Sandy nitrile palm provides outstanding grip in wet and oily conditions, hi-vis color improves awareness and compliance, and the brushed fleece liner creates additional warmth.
Certifications: ASTM F1790-97. ANSI Cut Level 3.
Ordering information: Gloves are sold by the dozen pairs.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty