Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
P-Vac Portable Vacuum System, Argos Technologies
Supplier: Argos Technologies
04397-07
10847-282EA 785.89 USD InStock
10847-282
P-Vac Portable Vacuum System, Argos Technologies
Pumps Vacuum Aspiration Systems
The P-Vac has been designed as a compact, flexible and portable liquid waste disposal system
The P-Vac can be operated with bottle top aspirators, or any glass or plastic bottles up to 2 liters. The unit is supplied with a quiet, adjustable diaphragm pump, which can empty a 2 liter container in just 40 seconds with an aspirating pipette. The hydrophobic filter protects the unit from over-filling.
The unit’s small footprint makes it ideal for the laboratory bench or laminar hood. It has been designed to accommodate a variety of collection vessels including bottle top aspirating filter units, plastic vacuum bottles, glass Erlenmeyer flasks, or any device that accepts vacuum. The rechargeable lithium ion battery allows for use where an inline vacuum source is not available, outside of the laboratory setting where a vacuum is required, or when the sample being aspirated is too dangerous for the house vacuum.
Ordering information: The P-Vac system is supplied with a .45um hydrophobic vacuum filter, silicone connection tubing, HandE-Vac hand operator with single channel pipette tip adapter and a universal main adapter.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty