Would you like to log in again?
Wide-Mouth Packers, Amber, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences
W216960 W216947 W217960 W216942 W216941 W216951 W216943 W216948 W216965 W216940 W216954 W216963 W216957 W217965 W216950 W216946 W216955 W216945 W217961 W216958 W216962 W216949 W216959 W216956 W216961 W217962 W217964 W217963 W216953 W216944 W216964 W216952 W216939
14235-892CS 98.94 USD InStock
14235-892 14235-894 14235-890 10847-552 14235-932 14235-910 14235-912 14235-934 14235-896 14235-898 14235-930 14235-908 14235-926 14235-904 14235-906 14235-928 10847-688 10847-686 10847-488 10847-486 14235-922 14235-900 14235-902 14235-924 14235-940 14235-942 14235-920 14235-918 14235-914 14235-936 14235-938 14235-916 10847-490
Wide-Mouth Packers, Amber, WHEATON®, DWK Life Sciences
Bottles Glass Bottles
Wheaton wide-mouth packers are manufactured from amber soda lime glass that conforms to USP Type III requirements.
Amber glass provides protection for light sensitive products. The wide-mouth design is ideal for use with dry powders. A partitioned carton protects bottles from scratches during shipping. Each case also includes a hazardous communication label and tamper evident seal if labeling and/or sealing is required.
Ordering information: Available with caps attached or in bulk packs without caps. Replacement screw caps can be purchased separately.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty