Would you like to log in again?
Straight Sided Bottles, Type III Glass, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
W217908 W217906 W217907 W217905
10847-522CS 105.76 USD InStock
10847-522 10847-520 10847-518 10847-516
Straight Sided Bottles, Type III Glass, WHEATON®, DWK Life Sciences
Bottles Glass Bottles
Bottles are made of Type III borosilicate glass.
  • Straight walls allow for complete removal of contents
  • Ideal for soil sampling and environmental applications
  • Supplied with attached screw caps
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty