Would you like to log in again?
Safety-Coated Reagent Bottles, Amber, Wide Mouth, DWK Life Sciences
Supplier: DWK Life Sciences
W217959 W217956 W217958 W217957
10847-568CS 82.14 USD InStock
10847-568 10847-566 10847-494 10847-492
Safety-Coated Reagent Bottles, Amber, Wide Mouth, DWK Life Sciences
Bottles Glass Bottles
These amber safety-coated bottles protect light-sensitive samples during storage or field applications.
  • Non-slip coating allows for greater ease and stability when handling wet or dry
  • Autoclaving/sterilizing bottle is not recommended
  • Bottles with caps are used for distilled water, analytical standards, and reagents
  • Metal foil liners are generally more resistant to organic solvents
  • Manufactured from soda lime glass
The strength, slip resistance, and impact absorption qualities of the plastisol safety coating reduce potential breakage and exposure to hazardous materials.
If a bottle is accidentally broken, the PVC based plastisol coating on the outside of bottle retains the contents long enough for proper disposal.
Certifications: Conforms to USP Type III requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty