Would you like to log in again?
WHEATON® Safety Coated Glass Bottles, DWK Life Sciences
Supplier: DWK Life Sciences
W217852 W217855 W217955 W217851 W217854 W217850 W217951 W217856 W217952 W217953 W217954 W217853
10847-558CS 179.06 USD InStock
10847-558 10847-578 10847-556 10847-564 10847-576 10847-496 10847-562 10847-574 10847-560 10847-572 10847-580 10847-570
WHEATON® Safety Coated Glass Bottles, DWK Life Sciences
Bottles Glass Bottles
Plastisol coats the exterior of the bottle to retain contents if a bottle is accidentally broken.
  • Made from clear or amber soda-lime glass that conforms to USP Type III requirements
  • Available with or without caps
  • 4, 8, 16, and 32 oz. Boston round style formats
These bottles are packaged in convenience packs with caps attached to maintain cleanliness.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty