Would you like to log in again?
Miriad Rapid HBc/HIV/HCV Antibody Test, LAB+ Format, MedMira
Supplier: MedMira
Miriad™
815311005978 815311005978
10847-592CS 22812.18 USD InStock
CA95062-809 10847-592
Miriad Rapid HBc/HIV/HCV Antibody Test, LAB+ Format, MedMira
Clinical Diagnostic Supplies Clinical Reagents and Kits
Miriad Rapid HBc/HIV/HCV Antibody Test employs MedMira Rapid Vertical Flow Technology to deliver the simultaneous diagnosis of Hepatitis B core, HIV-1/2, and Hepatitis C.
Miriad HBc/HIV/HCV features a built-in reagent control line, works with whole blood, serum and plasma specimens, and requires no additional equipment. Test kits can be stored for up to 18 months at 2 to 30 °C (35.6 to 86 °F).
In three easy steps users can obtain multiple instant results on a single test cartridge.
Ordering information: HBc/HIV/HCV Antibody Test Controls are available as an accessory to the test (10769-911).
Delivery information: Each box of Miriad HBc/HIV/HCV LAB+ Format includes 50 test kits, 50 sample prep tubes, 50 transfer pipettes and 2×30 ml bottles of universal buffer.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty