Would you like to log in again?
WHEATON® Packers, Wide Mouth, Round, HDPE, DWK Life Sciences
Supplier: DWK Life Sciences
W218821 W218822 W218823 W218825 W218826
10847-668CS 511.28 USD InStock
10847-668 10847-664 10847-662 10847-670 10847-660
WHEATON® Packers, Wide Mouth, Round, HDPE, DWK Life Sciences
Bottles Plastic Bottles
Wide mouth HDPE bottles with or without attached caps are available in a range of sizes.
  • High density polyethylene construction, natural color
  • Good chemical resistance
  • Wide mouth design is ideal for use with solids
Delivery information: Conveniently supplied with pre-attached white polypropylene screw caps with foamed polyethylene liners.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty