Would you like to log in again?
WHEATON® Standard Wide Mouth Bottle, Clear, DWK Life Sciences
Supplier: DWK Life Sciences
W217923
10847-680CS 77.27 USD InStock
10847-680
WHEATON® Standard Wide Mouth Bottle, Clear, DWK Life Sciences
Bottles Glass Bottles
Standard wide mouth bottles are made from high-quality clear type III soda-lime glass.
  • Largest bottles available
  • Without cap
Standard wide mouth bottles are perfect for general, small to large volume storage.
Ordering information: Caps for this series must be purchased separately.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty