Would you like to log in again?
Quintessential Stereotaxic Injector, Stoelting MS
Supplier: Stoelting
53311
10847-820EA 4502.27 USD InStock
10847-820
Quintessential Stereotaxic Injector, Stoelting MS
Syringe Dispensing Systems
The classic Quintessential Stereotaxic Injector (QSI™) sets the standard for automated injection providing versatility and control.
  • Preprogrammed syringe specifications
  • Out-of-range warnings
  • Accommodates Hamilton syringes sized 0.5 to 250 µl
This injector is used to control one or two independent injections with automatic infusion or withdrawal of nl and µl volumes.
Easy-to-use touch screen control box helps in program injection parameters such as volume, rate and syringe specifications.
Shipping dimensions: 12×12×10".
Ordering information: Steriotaxic sold separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty