Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
HayeSep® Packing Materials, Restek
Supplier: Restek
25047 25048 25037 25045 25054 25049 25050 25038 22560 25569 25573 25572 25053 25563 25570 25571 25575 25046 25574 25055 25033 25051 25039 25036 25032 25056 25568
10847-898EA 232.99 USD InStock
10847-898 10847-896 10847-852 10847-894 10847-872 10847-870 10847-858 10847-878 10848-038 10848-034 10848-014 10848-036 10847-880 10847-888 10847-866 10847-886 10847-884 10847-882 10847-860 10848-030 10848-032 10847-900 10847-868 10848-026 10848-028 10848-024 10847-890
HayeSep® Packing Materials, Restek
Chromatography Sorbents GC Sorbents
HayeSep® porous polymers are macroporous, spherical, ultrapure resins for performing unique separations in gas chromatography.
  • Thoroughly cleaned and preconditioned
  • No shrinkage
  • Low bleed
Columns packed with HayeSep® polymers produce lower bleed and more reproducible characteristics than those packed with any competing polymeric product, and they require minimal conditioning.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty