Would you like to log in again?
COLUMN CARBBK PKD 1 RT-1510 2.1MMX10FT
Supplier: Avantor
CarboBlack packed Column 10 ft^ CarboBlack packed Column, 1% Rt-1510, NOC, ID: 2.1 mm, Length: 10ft, OD: 1/8in, Material: Stainless Steel Tubing, Mesh: 60/80, Configuration: Agilent, Graphitized carbon black offers unique selectivity and very little adsorption for alcohol analyses, Siltek treated SS
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COLUMN CARBBK PKD 1 RT-1510 2.1MMX10FT 10847-958   Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty