Would you like to log in again?
COLUMN MICROPACKED GC HAYESEP Q 1MMX2M
Supplier: Avantor
Micropacked GC Column 2 m^ Micropacked GC Column, HayeSep Q, Inner Diameter: 1.00 mm, Length: 2 m, OD: 1/16in, Mesh: 100/120, Temperature: 275 deg C, Made from inert, flexible SilcoSmooth* tubing, with Braided Wire End Plugs, fit capillary injection systems, operate at flows exceeding 10 cc/min
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COLUMN MICROPACKED GC HAYESEP Q 1MMX2M 10848-104   Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty