Would you like to log in again?
4 CARBOWAX 20M 0.8 KOH CBKB 60/80
Supplier: Avantor
CarboBlack Packed Column 2m^ CarboBlack Packed Column 4% Carbowax 20M, On CarboBlack B, Inner Diameter: 2.0 mm, Length: 2 m, Material: SilcoSmooth Tubing, Mesh: 80/120, with Special Configuration, 0.8 KOH CarboBlack B support, with its higher surface area, Hold 10% nonsilicone liquid phase
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
4 CARBOWAX 20M 0.8 KOH CBKB 60/80 10848-144   Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty