Would you like to log in again?
10 RTX-5 SILCOPORT W 100/120 6FT 2MMID
Supplier: Avantor
Chromosorb* Diatomaceous Earth Column 6 ft^ Chromosorb* Diatomaceous Earth Column 10% Rtx-5, Inner Diameter: 2.1 mm, Length: 6 ft, Mesh: 100/120 Silcoport W, Material: Stainless Steel Tubing, Configuration: Agilent, bonded stationary phase packed, Low bleed levels, Longer column lifetimes, ABB 300
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
10 RTX-5 SILCOPORT W 100/120 6FT 2MMID 10848-422   Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty