Would you like to log in again?
PHASE 2A 2B 2C ON SILCOSTEEL TUBING
Supplier: Avantor
Refinery Gas Column^ Refinery Gas Column Set (3 set), phase: 2abc, finely tuned to resolve light hydrocarbons, will elute C5+ hydrocarbons ahead of C1 through C4 hydrocarbons, Material: Silico steel tubing, Configuration: Customer Supplied Dimensions
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
PHASE 2A 2B 2C ON SILCOSTEEL TUBING 10848-684   Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty