Would you like to log in again?
GCxGC Selectivity Kit (0.25 mm), Restek
Supplier: Restek
15131
10850-354EA 684.63 USD InStock
10850-354
GCxGC Selectivity Kit (0.25 mm), Restek
Chromatography Columns
GCxGC selectivity kits offer orthogonal separations through a wide range of stationary phases and accommodate varying sample capacities, speeds, and detectors.
  • Wide range of stationary phases offers orthogonal separations
  • High thermal stability increases system ruggedness
  • Unrivaled column inertness for accurate analysis of active compounds
  • 0.15, 0.18, or 0.25 mm Int.Ø formats accommodate varying sample capacities, speeds, and detectors
  • Secondary columns come in convenient 2 m lengths for economical methods development
Selectivity kits include secondary columns that come in convenient lengths for economical development of methods.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty