Would you like to log in again?
Rxi®-PAH Columns (Fused Silica), Restek
Supplier: Restek
49317 49316 49318
10850-562EA 769.04 USD InStock
10850-562 10059-574 10059-572
Rxi®-PAH Columns (Fused Silica), Restek
Chromatography Columns
The Rxi®-PAH GC columns were designed by Restek with a higher phenyl-content stationary phase that provides unique selectivity to separate important polycyclic aromatic hydrocarbons (PAH) for food safety that cannot be distinguished by mass spectrometry.
  • Ideal for EFSA PAH4 analysis—separates all priority compounds: benz[a]anthracene, chrysene, benzo[b]fluoranthene, and benzo[a]pyrene
  • Best resolution of chrysene from interfering PAHs, triphenylene, and cyclopenta[cd]pyrene
  • Complete separation of benzo [b], [k], [ j], and [a] fluoranthenes
  • 360°C thermal stability allows analysis of low-volatility dibenzo pyrenes
  • Mid-polarity proprietary phase
Even difficult priority compounds, such as the European Food Safety Authority (EFSA) PAH4, are easily separated and accurately quantified—results that cannot be achieved on typical GC columns. Arylene modification and surface bonding of the stationary phase increase thermal stability and ruggedness so relatively nonvolatile, higher molecular weight PAHs can be analyzed routinely without interference from column bleed. Excellent column efficiency means that the column can be trimmed for maintenance purposes many times without losing critical PAH separations, including those that are part of environmental methods, as well as food safety testing. The selectivity and efficiency of the Rxi®-PAH column make it ideal for EFSA PAH4 analysis; chrysene/triphenylene separation and resolution of all benzofluoranthenes are easily achieved.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty