Would you like to log in again?
COLUMN MXT 1HT SIM DIST 0.1UM 5MX0.53MM
Supplier: Avantor
MXT* 1HT Sim Dist Column 0.1um^ MXT* 1HT Sim Dist Column (Siltek* treated stainless steel), nonpolar phase; 100% dimethyl polysiloxane phase, Length: 5m, Internal Diameter: 0.53mm, df: 0.1um, Config: Coil on 11 Pin Cage, Temperature: -60 to 430 deg C, Lowest bleed for longest column lifetime
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COLUMN MXT 1HT SIM DIST 0.1UM 5MX0.53MM 10851-170   Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty