Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Corning® Gosselin™ Petri Dishes, Corning
Supplier: Corning
Corning®
BB64-02 SB93-102 BTX93-02 BT93-02 BP93B-101 BP93B-102 SB93-101 BB60-01 BP93D-102
10852-282CS 169.48 USD InStock
10852-282 10852-284 89238-018 10852-280 89200-954 89238-016 89200-956 10852-286 10852-178
Corning® Gosselin™ Petri Dishes, Corning
Culture Dishes Petri Dishes
Contact plates used in hygiene control within the pharmaceutical or food processing industry, as well as in hospital environments. Manufactured in a Class 100 environment with cleanroom packaging. Available aseptic or sterile (SAL 10⁻⁶).
  • Moulded grid on the bottom makes it possible to identify contamination per cm²
  • Grip lid ensures safety during transport
  • Domed base guarantees a saving in agar whilst maintaining safe sampling
  • Stable stacking
The Petri Dishes are packaged with easily accessible traceability features such as peelable labels with lot numbers, valid expiration date and sterilization information on each case. It is used in industrial microbiology labs such as Food and Beverage, Environmental/Water Testing, etc.
They are made of crystal polystyrene and are optimized for automation.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty