Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
HydraFlock® Dry Transport Systems, Puritan Medical Products
Supplier: Puritan Medical Products
HydraFlock®
25-3406-HBT 25-3706-HBT 25-36062HBT 253316HEMB80BT 25-33062HBT 25-3316-HBT 25-3318-HBT 25-3306-HBT 25-3317-HBT
10852-292BX 127.89 USD InStock
10852-292 10841-238 75788-576 10852-174 10852-294 10852-170 75791-994 10852-288 10852-166
HydraFlock® Dry Transport Systems, Puritan Medical Products
Swabs Transport Swabs
Get all your specimen collection devices in one place! Ideal for the collection and transport of specimen when no medium is required
Systems are designed for safe, sterile specimen transport. No matter which swab(s) are chosen, all transport systems include a round-bottom tube made from virgin polypropylene resin, which will securely hold the swab, maintaining a safe and sterile environment.
Tubes include a distinctive label describing product identity, expiry date, patient identification, and collection information. System applications include; specimen collection, rapid diagnostic testing, forensic evidence collection, DNA testing, and Buccal cell collection.
Choose from a range of HydraFlock® styles and tip sizes. Single or double options are available.
Certifications: Made in the USA. CE marked.
Ordering information: Swabs are supplied sterile. Double swabs are individually wrapped.
Packaging: There are 5 boxes per case, 100 individually wrapped dry transport tubes per box.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty