Would you like to log in again?
Tissue-Tek® FormaGO® Formalin Starter Kit, Sakura® Finetek
Supplier: Sakura
9150
10852-344KT 744.06 USD InStock
10852-344
Tissue-Tek® FormaGO® Formalin Starter Kit, Sakura® Finetek
Spill Control Materials
The Tissue-Tek® FormaGO® Formalin Starter Kit includes four (4) pouches of Formalin Neutralizer, one (1) jar of Formalin Absorption Granules, one (1) bag of 100 small Formalin Absorption Pads, one (1) bag of 20 medium Formalin Absorption Pads, one (1) bag of 10 large Formalin Absorption Pads, one (1) pack of 25 Formalin Absorption Wipes, and one (1) 9.5L Neutralizing Container.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty